tert-Butyl pyrrolidine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl pyrrolidine-1-carboxylate, pyrrolidine-1-carboxylic acid tert-butyl ester, N-BOC-Pyrrolidine

42582-0050 42582-0250
TS42582-0050EA 75 USD
TS42582-0050 TS42582-0250
tert-Butyl pyrrolidine-1-carboxylate 97%
tert-Butyl pyrrolidine-1-carboxylate
Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 80 °C (.2 mmHg)
Density: 0.977 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00216581
CAS Number: 86953-79-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR