5-Carboxyfluorescein 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42581-1000
42581-5000
TS42581-1000EA
107
USD
TS42581-1000
TS42581-5000
5-Carboxyfluorescein 95%
5-Carboxyfluorescein
Formula:
C₂₁H₁₂O₇.xH₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Specification Test Results
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Sulfated ash | ≤0.1 % |
Solubility | (in water) Sparingly soluble |
Related substances | Passes test |
Titration with NaOH | 99.5 to 100.5 % (On dried substance) |
Chloride (Cl) | ≤200 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...