4-Methyl-1H-imidazole-5-carbaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-5-imidazolecarboxaldehyde

42577-0050 42577-0250
TS42577-0050EA 74 USD
TS42577-0050 TS42577-0250
4-Methyl-1H-imidazole-5-carbaldehyde 99%
4-Methyl-1H-imidazole-5-carbaldehyde
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00173728
CAS Number: 68282-53-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 170 °C
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR