cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

42574-0010 42574-0050
TS42574-0010EA 59 USD
TS42574-0010 TS42574-0050
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00630580
CAS Number: 2746-19-2

Order Now

Specification Test Results

Appearance White to tan Solid
Infrared spectrum Conforms
Melting point 140 to 145 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR