Methyl 4-(1-hydroxyethyl)benzoate 90% tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-[4-(Methoxycarbonyl)phenyl]ethan-1-ol, 4-(Methoxycarbonyl)-α-methylbenzyl alcohol

42475-0010 42475-0050
TS42475-0010EA 91 USD
TS42475-0010 TS42475-0050
Methyl 4-(1-hydroxyethyl)benzoate 90% tech.
Methyl 4-(1-hydroxyethyl)benzoate
Formula: C₁₀H₁₂O₃
MW: 180.203 g/mol
Boiling Pt: 148…152 °C (1 mmHg)
Flash Pt: >93 °C
Storage Temperature: Ambient
MDL Number: MFCD00216476
CAS Number: 79322-76-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5360 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR