2-Thiophenesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thiophene-2-sulphonyl chloride

42089-0050 42089-0250
TS42089-0050EA 70 USD
TS42089-0050 TS42089-0250
2-Thiophenesulphonyl chloride 98%
2-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
MW: 182.65
Boiling Pt: 80 °C (1 mmHg)
Melting Pt: 32
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005426
CAS Number: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to blue-green to brown-purple Low melting solid or liquid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (Ex Cl)
GC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR