(±)-Tetrahydrofurfuryl alcohol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA

42044-0010 42044-0025 42044-2500
TS42044-0010EA 61 USD
TS42044-0010 TS42044-0025 TS42044-2500
(±)-Tetrahydrofurfuryl alcohol 99+%
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
Boiling Pt: 178 °C (760 mmHg)
Melting Pt: –80 °C
Density: 1.0543 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.0 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR