2-Quinolinol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Quinolin-2(1H)-one, Quinolin-2-ol, Carbostyril, 2-Quinolone, 2-Hydroxyquinoline

41881-0010 41881-0100
TS41881-0010EA 45 USD
TS41881-0010 TS41881-0100
2-Quinolinol 98%
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835

Order Now

Specification Test Results

Appearance White to brown-purple to purple Powder
Infrared spectrum Conforms
Melting point 194 to 200 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR