4-Fluoro-7-nitrobenzofurazan 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F
41545-0050
41545-0250
TS41545-0050EA
165
USD
TS41545-0050
TS41545-0250
4-Fluoro-7-nitrobenzofurazan 98%
4-Fluoro-7-nitrobenzofurazan
Formula:
C₆H₂FN₃O₃ MW: 183.1 g/mol Melting Pt: 52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010196 CAS Number: 29270-56-2 |
Specification Test Results
Appearance | Brown Crystalline chunks |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...