4-Methylphthalic anhydride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41479-0050
41479-0250
41479-1000
TS41479-0050EA
24
USD
TS41479-0050
TS41479-0250
TS41479-1000
4-Methylphthalic anhydride 96%
4-Methylphthalic anhydride
Formula:
C₉H₆O₃ Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041856 CAS Number: 19438-61-0 EINECS: 243-073-6 |
Specification Test Results
Appearance | White to yellow to light orange Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 88 to 93 °C |
GC | ≥95 % |
Titration by coupling | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...