ɑ-Methylene-γ-butyrolactone 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning

41426-0010 41426-0050
TS41426-0010EA 68 USD
TS41426-0010 TS41426-0050
ɑ-Methylene-γ-butyrolactone 95% stabilized
alpha-Methylene-gamma-butyrolactone
Formula: C₅H₆O₂
MW: 98.10 g/mol
Boiling Pt: 88…92 °C (12 mmHg)
Density: 1.12 g/cm³
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005407
CAS Number: 547-65-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4750 (20°C, 589 nm)
GC ≥94 %
Stabilizer 2,6-di-tert.-Butyl-p-cresol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR