Itaconic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione
41294-0250
41294-1000
TS41294-0250EA
49
USD
TS41294-0250
TS41294-1000
Itaconic anhydride 98%
Itaconic anhydride
Formula:
C₅H₄O₃ Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005530 CAS Number: 2170-03-8 EINECS: 218-518-2 |
Specification Test Results
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 66 to 70 °C |
Assay | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...