5-Chlorothiophene-2-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-thiophenecarboxaldehyde, 2-Chloro-5-formylthiophene

40508-0010 40508-0050
TS40508-0010EA 49 USD
TS40508-0010 TS40508-0050
5-Chlorothiophene-2-carboxaldehyde 97%
5-Chlorothiophene-2-carboxaldehyde
Formula: C₅H₃ClOS
MW: 146.59 g/mol
Boiling Pt: 94 °C (18 mmHg)
Density: 1.376 g/cm³
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047090
CAS Number: 7283-96-7

Order Now

Specification Test Results

Appearance Clear yellow to orange-red Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6040 to 1.6070 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR