2-Chlorobenzoxazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-1,3-benzoxazole

40453-0100 40453-0500
TS40453-0100EA 106 USD
TS40453-0100 TS40453-0500
2-Chlorobenzoxazole 98%
2-Chlorobenzoxazole
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.35 g/cm³
Flash Pt: 85 °C
MDL Number: MFCD00005766
CAS Number: 615-18-9
EINECS: 210-414-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5680 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR