Azobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

40154-0500 40154-2500
TS40154-0500EA 119 USD
TS40154-0500 TS40154-2500
Azobenzene 98%
Azobenzene
Formula: C₁₂H₁₀N₂
MW: 182.22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 66…69 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003022
CAS Number: 103-33-3
EINECS: 203-102-5
UN: 3077
ADR: 9,III
Merck Index: 12,00948

Order Now

Specification Test Results

Appearance Orange to red Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 65 to 69 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR