1-(2-Aminoethyl)piperazine 99%

Supplier: Thermo Scientific Chemicals
Danger

40076-0025 40076-1000 40076-5000
TS40076-0025EA 242 USD
TS40076-0025 TS40076-1000 TS40076-5000
1-(2-Aminoethyl)piperazine 99%
1-(2-Aminoethyl)piperazine
Formula: C₆H₁₅N₃
MW: 129.21 g/mol
Melting Pt: –19 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00005971
CAS Number: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5010 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR