9-Acridone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone
40025-0100
40025-0250
TS40025-0100EA
172
USD
TS40025-0100
TS40025-0250
9-Acridone 99%
9-Acridone
Formula:
C₁₃H₉NO Melting Pt: 360…362 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005019 CAS Number: 578-95-0 EINECS: 209-434-7 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with TBAH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...