2-(3-Bromopropoxy)tetrahydropyran 98% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3-Bromopropoxy)tetrahydro-2H-pyran

Stabilization: Stabilised with potassium carbonate

39973-0010 39973-0050
TS39973-0010EA 53 USD
TS39973-0010 TS39973-0050
2-(3-Bromopropoxy)tetrahydropyran 98% stabilised
2-(3-Bromopropoxy)tetrahydropyran
Formula: C₈H₁₅BrO₂
Boiling Pt: 64.5 °C (.8 mmHg)
Density: 1.314 g/cm³
Flash Pt: 106 °C
Storage Temperature: Freezer
MDL Number: MFCD00058593
CAS Number: 33821-94-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4790 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer K2CO3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR