4,5-Dichloro-2-methyl-2H-pyridazin-3-one 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5-Dichloro-2-methylpyridazin-3(2H)-one

39971-0010 39971-0050
TS39971-0010EA 40 USD
TS39971-0010 TS39971-0050
4,5-Dichloro-2-methyl-2H-pyridazin-3-one 97%
4,5-Dichloro-2-methyl-2H-pyridazin-3-one
Formula: C₅H₄Cl₂N₂O
MW: 179.01 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00051686
CAS Number: 933-76-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to yellow to brown Powder or crystalline powder or solid
Infrared spectrum Conforms
Melting point 87 to 92 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR