3-Chloro-2,4(3H,5H)-furandione 97%

Supplier: Thermo Scientific Chemicals
Warning

39965-0100 39965-0500
TS39965-0100EA 41 USD
TS39965-0100 TS39965-0500
3-Chloro-2,4(3H,5H)-furandione 97%
3-Chloro-2,4(3H,5H)-furandione
Formula: C₄H₃ClO₃
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00134273
CAS Number: 4971-55-5

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR