2,4,6-Trichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

39959-0010 39959-0050
TS39959-0010EA 64 USD
TS39959-0010 TS39959-0050
2,4,6-Trichloropyridine 98%
2,4,6-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Melting Pt: 33…37 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00971928
CAS Number: 16063-69-7

Order Now

Specification Test Results

Appearance White to light yellow Crystals or crystalline solid
Infrared spectrum Conforms
Melting point 31 to 37 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR