4-Amino-2,6-dichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

39870-0010 39870-0050
TS39870-0010EA 26 USD
TS39870-0010 TS39870-0050
4-Amino-2,6-dichloropyridine 98%
4-Amino-2,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00052832
CAS Number: 2587-02-2

Order Now

Specification Test Results

Appearance White to yellow-brown Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR