5-Bromo-2-nitropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

39864-0010 39864-0050
TS39864-0010EA 32 USD
TS39864-0010 TS39864-0050
5-Bromo-2-nitropyridine 99%
5-Bromo-2-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00160411
CAS Number: 39856-50-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 152 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR