6-Chloronicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-cyanopyridine, 6-Chloro-3-pyridinecarbonitrile, 2-Chloropyridine-5-carbonitrile

39839-0050 39839-0250
TS39839-0050EA 48 USD
TS39839-0050 TS39839-0250
6-Chloronicotinonitrile 97%
6-Chloronicotinonitrile
Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Boiling Pt: 107 °C (1 mmHg)
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00084941
CAS Number: 33252-28-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR