Isocytosine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4(1H)-pyrimidinone, 2-Aminopyrimidin-4(1H)-one

39832-0050 39832-0250
TS39832-0050EA 88 USD
TS39832-0050 TS39832-0250
Isocytosine 99%
Isocytosine
Formula: C₄H₅N₃O
MW: 111.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057557
CAS Number: 108-53-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Solubility (50 mg/ml in DMSO) Clear solution
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR