(R)-(+)-1-Boc-3-aminopyrrolidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine
39827-0010
39827-0050
TS39827-0010EA
113
USD
TS39827-0010
TS39827-0050
(R)-(+)-1-Boc-3-aminopyrrolidine 97%
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula:
C₉H₁₈N₂O₂ MW: 186.25 g/mol Flash Pt: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03419272 CAS Number: 147081-49-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | +2° to +4° (20°C, 589 nm) (c=1 in CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...