Piperidin-3-amine dihydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

39811-0010 39811-2500
TS39811-0010EA 519 USD
TS39811-0010 TS39811-2500
Piperidin-3-amine dihydrochloride 97%
Piperidin-3-amine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.081 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012773
CAS Number: 138060-07-8

Order Now

Specification Test Results

Appearance Off-white to beige Powder or solid
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR