3,4-Dihydroquinolin-2(1H)-one 98%

Supplier: Thermo Scientific Chemicals
Warning

39801-0010 39801-0100
TS39801-0010EA 48 USD
TS39801-0010 TS39801-0100
3,4-Dihydroquinolin-2(1H)-one 98%
3,4-Dihydroquinolin-2(1H)-one
Formula: C9H9NO
MW: 147.18 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00016722
CAS Number: 553-03-7

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 168 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR