2-Chloroisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

39779-0050 39779-0250
TS39779-0050EA 67 USD
TS39779-0050 TS39779-0250
2-Chloroisonicotinic acid 97%
2-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00191402
CAS Number: 6313-54-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR