1-Boc-4-(hydroxymethyl)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

39772-0010 39772-0050
TS39772-0010EA 17 USD
TS39772-0010 TS39772-0050
1-Boc-4-(hydroxymethyl)piperidine 97%
1-Boc-4-(hydroxymethyl)piperidine
Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD02094488
CAS Number: 123855-51-6

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR