4-Aminoimidazole-5-carboxamide 95%

Supplier: Thermo Scientific Chemicals
Warning

39755-0050 39755-0250
TS39755-0050EA 86 USD
TS39755-0050 TS39755-0250
4-Aminoimidazole-5-carboxamide 95%
4-Aminoimidazole-5-carboxamide
Formula: C₄H₆N₄O
Melting Pt: 164…170 °C
Storage Temperature: Ambient
MDL Number: MFCD02181040
CAS Number: 360-97-4

Order Now

Specification Test Results

Appearance Grey to purple Powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR