2-Methylnicotinic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid

39732-0010 39732-0050 39732-0250
TS39732-0010EA 96 USD
TS39732-0010 TS39732-0050 TS39732-0250
2-Methylnicotinic acid 98+%
2-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 225…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00013449
CAS Number: 3222-56-8

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR