5-Formyl-2-thiopheneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

39715-0010 39715-0050
TS39715-0010EA 81 USD
TS39715-0010 TS39715-0050
5-Formyl-2-thiopheneboronic acid 97%
5-Formyl-2-thiopheneboronic acid
Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093666
CAS Number: 4347-33-5

Order Now

Specification Test Results

Appearance White to orange Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR