tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-BOC-3-pyrrolidinone

39705-0010 39705-0100
TS39705-0010EA 59 USD
TS39705-0010 TS39705-0100
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631194
CAS Number: 101385-93-7

Order Now

Specification Test Results

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR