1-Boc-3-piperidone 97%

Supplier: Thermo Scientific Chemicals
Warning

39700-0050 39700-0250 39700-1000
TS39700-0050EA 62 USD
TS39700-0050 TS39700-0250 TS39700-1000
1-Boc-3-piperidone 97%
1-Boc-3-piperidone
Formula: C₁₀H₁₇NO₃
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now

Specification Test Results

Appearance White to yellow to orange Solid or liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR