(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S,3S)-N-t-Boc-3-amino-1,2-epoxy-4-phenylbutane, (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, Darunavir Impurity 22

39502-0010 39502-0050 39502-0250
TS39502-0010EA 161 USD
TS39502-0010 TS39502-0050 TS39502-0250
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane
Formula: C₁₅H₂₁NO₃
MW: 263.34 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02258997
CAS Number: 98737-29-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR