2-Phenylthiazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39437-0010
39437-0050
TS39437-0010EA
59
USD
TS39437-0010
TS39437-0050
2-Phenylthiazole 98%
2-Phenylthiazole
Formula:
C₉H₇NS MW: 161.2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00272327 CAS Number: 1826-11-5 |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...