5-Bromopicolinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

39399-0010 39399-0050 39399-0250
TS39399-0010EA 28 USD
TS39399-0010 TS39399-0050 TS39399-0250
5-Bromopicolinic acid 97%
5-Bromopicolinic acid
Formula: C₆H₄BrNO₂
Melting Pt: 177…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234149
CAS Number: 30766-11-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR