2-Amino-5-methylthiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methylthiazol-2-ylamine

39279-0050 39279-0250 39279-1000
TS39279-0050EA 27 USD
TS39279-0050 TS39279-0250 TS39279-1000
2-Amino-5-methylthiazole 98%
2-Amino-5-methylthiazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00078317
CAS Number: 7305-71-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR