1-Bromoisoquinoline 95+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39259-0010
39259-0050
TS39259-0010EA
121
USD
TS39259-0010
TS39259-0050
1-Bromoisoquinoline 95+%
1-Bromoisoquinoline
Formula:
C₉H₆BrN MW: 208.0546 g/mol Melting Pt: 43…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234478 CAS Number: 1532-71-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow to brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 42 to 48 °C |
HPLC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...