3-Bromo-4-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-picoline

39232-0010 39232-0050
TS39232-0010EA 30 USD
TS39232-0010 TS39232-0050
3-Bromo-4-methylpyridine 97%
3-Bromo-4-methylpyridine
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 76 °C (10 mmHg)
Density: 1.549 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00082592
CAS Number: 3430-22-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5640 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR