5-Cyano-2-thiopheneboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

39127-0010 39127-0050
TS39127-0010EA 175 USD
TS39127-0010 TS39127-0050
5-Cyano-2-thiopheneboronic acid 98%
5-Cyano-2-thiopheneboronic acid
Formula: C₅H₄BNO₂S
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094029
CAS Number: 305832-67-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to pink Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR