9-Vinylcarbazole 97%

Supplier: Thermo Scientific Chemicals
Warning

38722-0050 38722-0250 38722-1000
TS38722-0050EA 29 USD
TS38722-0050 TS38722-0250 TS38722-1000
9-Vinylcarbazole 97%
9-Vinylcarbazole
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 154 °C (3 mmHg)
Melting Pt: 63…65 °C
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00004966
CAS Number: 1484-13-5
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Off-white to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR