4-Amino-1-boc-piperidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
38486-0010
38486-0050
TS38486-0010EA
25
USD
TS38486-0010
TS38486-0050
4-Amino-1-boc-piperidine 98%
4-Amino-1-Boc-piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01076201 CAS Number: 87120-72-7 UN: 3259 ADR: 8,III |
Specification Test Results
Appearance | White to light yellow to beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...