1-Allylpiperazine 99%

Supplier: Thermo Scientific Chemicals
Danger

38473-0010 38473-0050
TS38473-0010EA 30 USD
TS38473-0010 TS38473-0050
1-Allylpiperazine 99%
1-Allylpiperazine
Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 181…182 °C (760 mmHg)
Density: 1 g/cm³
Flash Pt: 51.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00167970
CAS Number: 13961-36-9
UN: 2734
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR