2,4-Dihydroxy-6-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

38428-0010 38428-0050
TS38428-0010EA 32 USD
TS38428-0010 TS38428-0050
2,4-Dihydroxy-6-methylpyridine 97%
2,4-Dihydroxy-6-methylpyridine
Formula: C₆H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD01860849
CAS Number: 3749-51-7

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR