1,3-Dichloroisoquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

38241-0010 38241-0050
TS38241-0010EA 88 USD
TS38241-0010 TS38241-0050
1,3-Dichloroisoquinoline 97%
1,3-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00034750
CAS Number: 7742-73-6

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR