Piperine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

38145-0050 38145-0250
TS38145-0050EA 45 USD
TS38145-0050 TS38145-0250
Piperine 98%
Piperine
Formula: C₁₇H₁₉NO₃
MW: 285.34 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00005839
CAS Number: 94-62-2

Order Now

Specification Test Results

Appearance Off-white to yellow to tan Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR