3-Bromo-4-chloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37964-0010
37964-0050
TS37964-0010EA
94
USD
TS37964-0010
TS37964-0050
3-Bromo-4-chloropyridine 98%
3-Bromo-4-chloropyridine
Formula:
C₅H₃BrClN |
MDL Number:
MFCD00234008 CAS Number: 36953-42-1 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid after melting |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...