5-Bromo-2-chloro-3-nitropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37957-0050
37957-0250
TS37957-0050EA
27
USD
TS37957-0050
TS37957-0250
5-Bromo-2-chloro-3-nitropyridine 98%
5-Bromo-2-chloro-3-nitropyridine
Formula:
C₅H₂BrClN₂O₂ Melting Pt: 64…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00222270 CAS Number: 67443-38-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 64 to 69 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...